Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015072.4 | NP_055887.3 | 19 | 1943 | ||
GenBank mRNA | AB023215.1 | - | 17 | 1503 | ||
AK309748.1 | - | 20 | 1925 | |||
AL136808.1 | - | 5 | 391 | |||
AY237126.1 | - | 19 | 1873 | |||
BC016625.1 | - | 6 | 524 | |||
BC136472.1 | - | 19 | 1833 | 9020461 | ||
BC136473.1 | - | 19 | 1833 | 9020462 | ||
BX647530.1 | - | 12 | 974 | |||
CR533554.1 | - | 4 | 321 |