Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171166.1 | NP_001164637.1 | 8 | 760 | ||
NM_001171167.1 | NP_001164638.1 | 9 | 1199 | |||
NM_001171168.1 | NP_001164639.1 | 8 | 1127 | |||
NM_015099.3 | NP_055914.2 | 9 | 855 | |||
XM_006721478.3 | XP_006721541.1 | 9 | 1502 | |||
XM_006721481.3 | XP_006721544.1 | 8 | 1415 | |||
XM_006721482.3 | XP_006721545.1 | 7 | 548 | |||
XM_011523746.2 | XP_011522048.1 | 9 | 862 | |||
XM_011523747.2 | XP_011522049.1 | 9 | 846 | |||
XM_011523748.2 | XP_011522050.1 | 9 | 861 | |||
XM_011523749.2 | XP_011522051.1 | 9 | 845 | |||
GenBank mRNA | AB020716.1 | - | 9 | 816 | ||
AK302528.1 | - | 9 | 794 | |||
AL831849.1 | - | 8 | 1127 | |||
AL833974.1 | - | 8 | 745 | |||
BC136534.1 | - | 8 | 784 | 9020524 | ||
BC142695.1 | - | 9 | 846 | 40148717 | ||
BC144232.1 | - | 7 | 712 | 9052752 | ||
BC167148.1 | - | 9 | 816 | 8862515 | ||
BC167149.1 | - | 9 | 816 | 8862516 | ||
BC167151.1 | - | 9 | 816 | 8862527 | ||
BC167160.1 | - | 9 | 816 | 8862594 |