Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303420.1 | NP_001290349.1 | 8 | 1528 | ||
NM_001303421.1 | NP_001290350.1 | 8 | 1042 | |||
NM_015101.3 | NP_055916.1 | 8 | 1528 | |||
XM_011509332.2 | XP_011507634.1 | 9 | 1334 | |||
XM_011509334.2 | XP_011507636.1 | 9 | 1141 | |||
XM_011509335.2 | XP_011507637.1 | 9 | 1192 | |||
XM_011509336.2 | XP_011507638.1 | 9 | 969 | |||
XM_011509337.2 | XP_011507639.1 | 5 | 1268 | |||
XM_011509338.2 | XP_011507640.1 | 6 | 1020 | |||
XM_017000750.1 | XP_016856239.1 | 8 | 1093 | |||
XM_017000751.1 | XP_016856240.1 | 3 | 952 | |||
GenBank mRNA | AB011156.1 | - | 8 | 1453 | ||
AF288389.1 | - | 8 | 1487 | |||
AK095090.1 | - | 3 | 763 | |||
AK095192.1 | - | 8 | 1016 | |||
AK293973.1 | - | 8 | 1235 | |||
BC035672.1 | - | 8 | 1510 | 5740842 |