Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303421.1 | NP_001290350.1 | 12 | 1936 | ||
NM_015101.3 | NP_055916.1 | 12 | 2422 | |||
XM_011509332.2 | XP_011507634.1 | 13 | 2228 | |||
XM_011509334.2 | XP_011507636.1 | 13 | 2035 | |||
XM_011509335.2 | XP_011507637.1 | 13 | 2086 | |||
XM_011509336.2 | XP_011507638.1 | 13 | 1863 | |||
XM_011509337.2 | XP_011507639.1 | 9 | 2162 | |||
XM_011509338.2 | XP_011507640.1 | 10 | 1914 | |||
XM_017000750.1 | XP_016856239.1 | 12 | 1987 | |||
XM_017000751.1 | XP_016856240.1 | 7 | 1846 | |||
GenBank mRNA | AB011156.1 | - | 12 | 2347 | ||
AF288389.1 | - | 12 | 2381 | |||
AK095090.1 | - | 7 | 1657 | |||
AK095192.1 | - | 12 | 1910 | |||
AK127287.1 | - | 4 | 1927 |