Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015103.2 | NP_055918.2 | 1 | 1107 | ||
XM_011512588.1 | XP_011510890.1 | 1 | 1022 | |||
XM_011512589.1 | XP_011510891.1 | 1 | 1022 | |||
XM_011512590.2 | XP_011510892.1 | 1 | 1022 | |||
GenBank mRNA | AB014520.2 | - | 1 | 1186 | ||
AY116661.1 | - | 1 | 1187 | |||
BC150280.1 | - | 1 | 1186 | 9007263 |