Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015103.2 | NP_055918.2 | 19 | 3741 | ||
XM_011512588.1 | XP_011510890.1 | 19 | 3656 | |||
XM_011512589.1 | XP_011510891.1 | 16 | 3266 | |||
XM_011512590.2 | XP_011510892.1 | 19 | 3656 | |||
XM_011512592.1 | XP_011510894.1 | 7 | 824 | |||
GenBank mRNA | AB014520.2 | - | 19 | 3820 | ||
AY116661.1 | - | 19 | 3821 | |||
BC150280.1 | - | 19 | 3820 | 9007263 |