Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK8IP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040439.1 | NP_001035529.1 | 12 | 1627 | ||
NM_001318852.1 | NP_001305781.1 | 13 | 1685 | |||
NM_015133.4 | NP_055948.2 | 13 | 1682 | |||
XM_006720869.2 | XP_006720932.1 | 13 | 3115 | |||
XM_011522429.2 | XP_011520731.1 | 14 | 1709 | |||
XM_011522430.2 | XP_011520732.1 | 13 | 1691 | |||
XM_011522433.2 | XP_011520735.1 | 9 | 2332 | |||
XM_017023079.1 | XP_016878568.1 | 12 | 1667 | |||
XM_017023081.1 | XP_016878570.1 | 12 | 1157 | |||
GenBank mRNA | AB028989.2 | - | 13 | 1645 | ||
BC137123.1 | - | 13 | 1597 | 9021120 | ||
BC137124.1 | - | 13 | 1597 | 9021121 | ||
BC144486.1 | - | 13 | 1600 | 9053013 | ||
BC150266.1 | - | 13 | 1645 | 9007222 |