Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANK1 SIRNA Assays ›
Gene Symbol:
KANK1
Gene Name:
KN motif and ankyrin repeat domains 1
Gene Aliases:
ANKRD15, CPSQ2, KANK
Chromosome Location:
Chr.9: 470294 - 746106 on Build GRCh38
UniGene ID:
Hs.306764
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136799
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256876.1
NP_001243805.1
16
5407
NM_001256877.1
NP_001243806.1
13
5516
NM_015158.3
NP_055973.2
12
4901
NM_153186.4
NP_694856.1
11
5163
XM_005251411.2
XP_005251468.1
11
5484
XM_005251414.3
XP_005251471.1
10
5947
XM_005251415.2
XP_005251472.1
10
5676
XM_005251416.2
XP_005251473.1
10
5452
XM_005251417.3
XP_005251474.1
10
5943
XM_005251418.2
XP_005251475.1
10
5954
XM_005251419.2
XP_005251476.1
10
5645
XM_006716743.2
XP_006716806.1
11
5460
XM_011517819.1
XP_011516121.1
12
5575
XM_011517820.1
XP_011516122.1
11
5592
XM_011517821.1
XP_011516123.1
11
5519
XM_017014511.1
XP_016870000.1
12
5697
XM_017014512.1
XP_016870001.1
11
7008
XM_017014513.1
XP_016870002.1
13
6210
XM_017014514.1
XP_016870003.1
12
5313
XM_017014515.1
XP_016870004.1
13
5049
XM_017014516.1
XP_016870005.1
11
5842
XM_017014517.1
XP_016870006.1
12
5259
XM_017014518.1
XP_016870007.1
12
4847
XM_017014519.1
XP_016870008.1
10
5656
XM_017014520.1
XP_016870009.1
10
5244
XM_017014524.1
XP_016870013.1
10
5602
XM_017014525.1
XP_016870014.1
11
5019
XM_017014526.1
XP_016870015.1
11
5929
XM_017014527.1
XP_016870016.1
11
5093
XM_017014528.1
XP_016870017.1
12
4947
XM_017014529.1
XP_016870018.1
11
5360
XM_017014530.1
XP_016870019.1
11
4869
XM_017014531.1
XP_016870020.1
10
5622
XM_017014532.1
XP_016870021.1
10
5896
XM_017014533.1
XP_016870022.1
10
5393
XM_017014534.1
XP_016870023.1
11
5039
XM_017014535.1
XP_016870024.1
11
5305
XM_017014536.1
XP_016870025.1
11
5008
XM_017014537.1
XP_016870026.1
9
5225
XM_017014538.1
XP_016870027.1
9
5710
XM_017014542.1
XP_016870031.1
10
4575
GenBank mRNA
AK025428.1
-
10
3046
AK025636.1
-
5
2098
AL833015.1
-
8
1813
AL833161.1
-
12
4881
BC020040.1
-
10
3055
BC037495.2
-
16
5410
5265517
BC038116.2
-
11
5073
5261060
BX648931.1
-
11
7027
D79994.2
-
12
5466
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of neuron projection development
positive regulation of Wnt signaling pathway
negative regulation of cell migration
negative regulation of actin filament polymerization
negative regulation of Rho protein signal transduction
positive regulation of catenin import into nucleus
negative regulation of insulin receptor signaling pathway
positive regulation of wound healing
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of ruffle assembly
regulation of establishment of cell polarity
negative regulation of lamellipodium morphogenesis
protein binding
beta-catenin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-8pgsz:80/100.66.79.163:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline