Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003816.2 | NP_003807.1 | 3 | 299 | ||
NR_027638.1 | 3 | 299 | ||||
NR_027639.1 | 3 | 299 | ||||
NR_027878.1 | 3 | 299 | ||||
XM_011544682.2 | XP_011542984.1 | 3 | 549 | |||
XM_017013942.1 | XP_016869431.1 | 3 | 242 | |||
GenBank mRNA | AF495383.1 | - | 3 | 254 | ||
BC126406.1 | - | 3 | 259 | 8992122 | ||
BC143923.1 | - | 3 | 259 | 9052441 | ||
BC143924.1 | - | 3 | 259 | |||
U41766.1 | - | 3 | 299 |