Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBC1D1 SIRNA Assays ›
Gene Symbol:
TBC1D1
Gene Name:
TBC1 domain family member 1
Gene Aliases:
TBC, TBC1
Chromosome Location:
Chr.4: 37891084 - 38139175 on Build GRCh38
UniGene ID:
Hs.176503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136811
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBC1D1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253912.1
NP_001240841.1
21
4106
NM_001253913.1
NP_001240842.1
6
1772
NM_001253914.1
NP_001240843.1
6
1418
NM_001253915.1
NP_001240844.1
4
968
NM_015173.3
NP_055988.2
20
4133
XM_005262646.2
XP_005262703.1
23
4454
XM_005262647.2
XP_005262704.1
22
4415
XM_005262649.2
XP_005262706.1
22
3907
XM_006714001.2
XP_006714064.1
11
2234
XM_011513659.2
XP_011511961.1
24
4514
XM_011513660.2
XP_011511962.1
23
4475
XM_011513662.2
XP_011511964.1
23
4394
XM_011513663.2
XP_011511965.1
22
4355
XM_011513664.2
XP_011511966.1
23
4352
XM_011513665.2
XP_011511967.1
22
4232
XM_011513666.2
XP_011511968.1
21
4193
XM_011513670.2
XP_011511972.1
9
2236
XM_017007918.1
XP_016863407.1
22
4313
XM_017007919.1
XP_016863408.1
21
4295
XM_017007920.1
XP_016863409.1
21
4172
XM_017007921.1
XP_016863410.1
19
3448
GenBank mRNA
AB029031.1
-
14
2560
AK027355.1
-
12
2332
AK057182.1
-
6
1772
AK074954.1
-
6
1417
AK295746.1
-
21
3929
BC014529.2
-
18
2971
BC050321.1
-
20
4130
5262043
BC053648.1
-
18
2946
BC126979.1
-
10
2248
CR996632.1
-
1
450
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
regulation of vesicle fusion
regulation of protein localization
membrane organization
activation of GTPase activity
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-f7mcl:80/100.66.77.65:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline