Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145114.1 | NP_001138586.1 | 13 | 1698 | ||
NM_001284337.1 | NP_001271266.1 | 12 | 1581 | |||
NM_015179.3 | NP_055994.2 | 15 | 1881 | |||
XM_011539555.1 | XP_011537857.1 | 14 | 1750 | |||
XM_011539556.2 | XP_011537858.1 | 15 | 1851 | |||
GenBank mRNA | AB014590.1 | - | 12 | 1494 | ||
AK000238.1 | - | 12 | 1291 | |||
AK021460.1 | - | 380 | ||||
AK022496.1 | - | 15 | 1852 | |||
AK131283.1 | - | 9 | 957 | |||
AK296315.1 | - | 13 | 1562 | |||
BC012745.2 | - | 15 | 1819 | 4025242 |