Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE2 SIRNA Assays ›
Gene Symbol:
SYNE2
Gene Name:
spectrin repeat containing nuclear envelope protein 2
Gene Aliases:
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location:
Chr.14: 63761904 - 64226449 on Build GRCh38
UniGene ID:
Hs.745014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136819
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015180.4
NP_055995.4
110
20042
NM_182910.2
NP_878914.1
4
773
NM_182913.2
NP_878917.1
6
878
NM_182914.2
NP_878918.2
111
20111
XM_005267454.1
XP_005267511.1
111
20111
XM_005267456.1
XP_005267513.1
111
20111
XM_005267457.1
XP_005267514.1
111
20111
XM_005267458.1
XP_005267515.1
110
20042
XM_005267459.1
XP_005267516.1
110
20042
XM_011536574.1
XP_011534876.1
111
20153
XM_011536575.2
XP_011534877.1
111
20130
XM_011536576.2
XP_011534878.1
112
20310
XM_011536577.2
XP_011534879.1
111
20077
XM_011536578.1
XP_011534880.1
111
20153
XM_011536579.1
XP_011534881.1
111
20153
XM_011536580.1
XP_011534882.1
111
20153
XM_011536581.1
XP_011534883.1
110
20084
XM_011536582.1
XP_011534884.1
110
20036
XM_017021101.1
XP_016876590.1
111
20155
XM_017021102.1
XP_016876591.1
110
20198
XM_017021103.1
XP_016876592.1
13
2190
XM_017021104.1
XP_016876593.1
13
2190
GenBank mRNA
AB023228.3
-
62
9783
AF357236.1
-
110
19581
AF435011.1
-
110
20024
AF495911.1
-
110
20009
AK074055.1
-
22
3509
AK090430.1
-
28
4418
AK297874.1
-
6
839
AK298465.1
-
5
553
AL080133.1
-
16
2555
AL117404.1
-
4
1295
AY061757.1
-
11
1646
AY061758.1
-
6
875
AY061759.1
-
58
9345
AY184204.1
-
4
1295
AY184205.1
-
9
1527
BX537642.1
-
31
4856
BX538095.1
-
19
2976
BX640974.1
-
12
1857
CR749324.1
-
31
4974
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nuclear envelope organization
nuclear migration
establishment or maintenance of cell polarity
fibroblast migration
positive regulation of cell migration
nuclear migration along microfilament
protein localization to nucleus
centrosome localization
cytoskeletal anchoring at nuclear membrane
actin binding
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0