Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE2 SIRNA Assays ›
Gene Symbol:
SYNE2
Gene Name:
spectrin repeat containing nuclear envelope protein 2
Gene Aliases:
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location:
Chr.14: 63761904 - 64226449 on Build GRCh38
UniGene ID:
Hs.745014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136820
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015180.4
NP_055995.4
112
20390
NM_182910.2
NP_878914.1
6
1163
NM_182913.2
NP_878917.1
8
1268
NM_182914.2
NP_878918.2
113
20459
XM_005267454.1
XP_005267511.1
113
20501
XM_005267456.1
XP_005267513.1
113
20501
XM_005267457.1
XP_005267514.1
113
20459
XM_005267458.1
XP_005267515.1
112
20432
XM_005267459.1
XP_005267516.1
112
20390
XM_011536574.1
XP_011534876.1
113
20543
XM_011536575.2
XP_011534877.1
113
20520
XM_011536576.2
XP_011534878.1
114
20700
XM_011536577.2
XP_011534879.1
113
20467
XM_011536578.1
XP_011534880.1
113
20543
XM_011536579.1
XP_011534881.1
113
20501
XM_011536580.1
XP_011534882.1
113
20501
XM_011536581.1
XP_011534883.1
112
20474
XM_011536582.1
XP_011534884.1
112
20426
XM_017021101.1
XP_016876590.1
113
20545
XM_017021102.1
XP_016876591.1
112
20588
XM_017021103.1
XP_016876592.1
15
2580
XM_017021104.1
XP_016876593.1
15
2580
GenBank mRNA
AB023228.3
-
64
10131
AF357236.1
-
112
19929
AF435011.1
-
112
20372
AF495911.1
-
112
20357
AK001876.1
-
3
302
AK074055.1
-
24
3857
AK090430.1
-
30
4766
AK297874.1
-
8
1187
AK298465.1
-
7
901
AL080133.1
-
18
2903
AL117404.1
-
6
1685
AY061757.1
-
13
1994
AY061758.1
-
8
1265
AY061759.1
-
60
9693
AY184204.1
-
6
1685
AY184205.1
-
11
1875
BX537642.1
-
33
5204
BX538095.1
-
21
3366
BX640974.1
-
14
2205
CR749324.1
-
33
5364
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nuclear envelope organization
nuclear migration
establishment or maintenance of cell polarity
fibroblast migration
positive regulation of cell migration
nuclear migration along microfilament
protein localization to nucleus
centrosome localization
cytoskeletal anchoring at nuclear membrane
actin binding
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0