Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015201.4 | NP_056016.1 | 14 | 1992 | ||
GenBank mRNA | AB060694.1 | - | 13 | 1660 | ||
AK290677.1 | - | 14 | 1954 | |||
AK293719.1 | - | 13 | 1636 | |||
AK310015.1 | - | 12 | 1566 | |||
AK315612.1 | - | 14 | 1961 | |||
BC001086.2 | - | 13 | 1718 | |||
BC005160.2 | - | 13 | 1745 | |||
BC013787.1 | - | 14 | 1954 | 3872212 | ||
BC013980.2 | - | 14 | 1957 | 3633864 | ||
BC017674.2 | - | 14 | 1944 | 3905068 | ||
BC073813.1 | - | 9 | 1251 | |||
BC098446.1 | - | 10 | 1388 | |||
D50914.1 | - | 13 | 1732 |