Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015323.4 | NP_056138.1 | 6 | 533 | ||
GenBank mRNA | AB018319.1 | - | 6 | 502 | ||
AK295934.1 | - | 6 | 487 | |||
AK307651.1 | - | 6 | 518 | |||
AK316453.1 | - | 6 | 785 | |||
BC015377.1 | - | 6 | 520 | |||
BC028608.1 | - | 6 | 512 | 4825054 | ||
BC036379.2 | - | 6 | 518 | 5267922 | ||
BP337693.1 | - | 6 | 533 |