Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282531.1 | NP_001269460.1 | 6 | 1294 | ||
NM_001282532.1 | NP_001269461.1 | 4 | 1162 | |||
NM_015339.3 | NP_056154.1 | 5 | 1210 | |||
NM_181442.2 | NP_852107.1 | 4 | 1035 | |||
XM_011528747.2 | XP_011527049.1 | 4 | 1387 | |||
XM_011528748.2 | XP_011527050.1 | 4 | 944 | |||
XM_017027757.1 | XP_016883246.1 | 5 | 5113 | |||
XM_017027758.1 | XP_016883247.1 | 4 | 2100 | |||
XM_017027759.1 | XP_016883248.1 | 4 | 970 | |||
GenBank mRNA | AB018327.1 | - | 3 | 779 | ||
AF250860.1 | - | 5 | 1210 | |||
AK314734.1 | - | 4 | 949 | |||
AL080163.1 | - | 1 | 574 | |||
BC075794.1 | - | 5 | 1433 | 30342099 | ||
BC090933.1 | - | 4 | 1162 | 3066721 |