Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016272.3 | NP_057356.1 | 2 | 2991 | ||
XM_005261315.2 | XP_005261372.1 | 2 | 4202 | |||
XM_006724105.3 | XP_006724168.1 | 2 | 3508 | |||
XM_017028539.1 | XP_016884028.1 | 2 | 5284 | |||
GenBank mRNA | AB051450.1 | - | 2 | 2750 | ||
BC038957.1 | - | 2 | 2764 | 5786640 |