Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320546.1 | NP_001307475.1 | 2 | 1387 | ||
NM_001320547.1 | NP_001307476.1 | 2 | 758 | |||
NM_018031.4 | NP_060501.3 | 2 | 1173 | |||
GenBank mRNA | AF099100.1 | - | 2 | 842 | ||
AK225653.1 | - | 2 | 836 | |||
AK295145.1 | - | 2 | 1173 | |||
AK296387.1 | - | 2 | 458 | |||
AK304117.1 | - | 2 | 758 | |||
AK309913.1 | - | 2 | 1053 | |||
AL833910.1 | - | 1 | 19 | |||
BC101707.1 | - | 2 | 832 | 8069213 | ||
BC113467.1 | - | 2 | 832 | 8322519 | ||
BC143486.1 | - | 2 | 832 |