Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMG20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304504.1 | NP_001291433.1 | 7 | 993 | ||
NM_001304505.1 | NP_001291434.1 | 7 | 905 | |||
NM_018200.3 | NP_060670.1 | 8 | 1086 | |||
XM_011521158.2 | XP_011519460.1 | 7 | 949 | |||
GenBank mRNA | AF146222.1 | - | 7 | 726 | ||
AK001601.1 | - | 7 | 756 | |||
AK223100.1 | - | 7 | 752 | |||
AK297449.1 | - | 6 | 686 | |||
AK316281.1 | - | 4 | 361 | |||
AL355737.1 | - | 6 | 648 | |||
BC021959.1 | - | 8 | 836 | 3908842 | ||
BT006716.1 | - | 6 | 635 |