Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021148.2 | NP_066971.2 | 4 | 1978 | ||
NR_003099.1 | 5 | 2067 | ||||
GenBank mRNA | AK090648.1 | - | 8 | 3114 | ||
AL832810.1 | - | 1 | 1670 | |||
BC063818.1 | - | 5 | 2062 | 6503687 | ||
X78932.1 | - | 1 | 967 |