Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM175B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032182.3 | NP_115558.3 | 9 | 842 | ||
XM_011539554.1 | XP_011537856.1 | 8 | 746 | |||
XM_017015986.1 | XP_016871475.1 | 7 | 720 | |||
GenBank mRNA | AK025991.1 | - | 2 | 124 | ||
AK296677.1 | - | 9 | 814 | |||
AL537809.3 | - | 9 | 794 | |||
BC008999.1 | - | 9 | 804 | 2900632 | ||
D63877.1 | - | 9 | 811 |