Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF159165.1 | - | 1 | 788 | ||
AK023820.1 | - | 8 | 1573 | |||
AK127944.1 | - | 3 | 2116 |