Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015286.5 | NP_056101.5 | 4 | 1585 | ||
NM_145728.2 | NP_663780.2 | 4 | 1585 | |||
XM_017022035.1 | XP_016877524.1 | 4 | 1596 | |||
GenBank mRNA | AB002351.2 | - | 4 | 1623 | ||
AF359284.1 | - | 4 | 1576 | |||
AJ310521.1 | - | 4 | 1579 | |||
AJ310522.1 | - | 4 | 1579 | |||
AL833262.1 | - | 4 | 1076 | |||
BC151243.1 | - | 4 | 1630 | 8860516 |