Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007216.3 | NP_009147.3 | 2 | 245 | ||
NM_181507.1 | NP_852608.1 | 3 | 402 | |||
NM_181508.1 | NP_852609.1 | 2 | 58 | |||
XM_011519862.1 | XP_011518164.1 | 3 | 393 | |||
XM_011519863.1 | XP_011518165.1 | 3 | 301 | |||
XM_011519864.1 | XP_011518166.1 | 3 | 233 | |||
XM_011519866.1 | XP_011518168.1 | 2 | 266 | |||
XM_011519867.1 | XP_011518169.1 | 2 | 132 | |||
XM_011519868.2 | XP_011518170.1 | 2 | 373 | |||
XM_017017149.1 | XP_016872638.1 | 3 | 301 | |||
XM_017017150.1 | XP_016872639.1 | 3 | 233 | |||
XM_017017154.1 | XP_016872643.1 | 2 | 132 | |||
GenBank mRNA | AB023234.1 | - | 2 | 58 | ||
AF534400.1 | - | 2 | 58 | |||
AF534401.1 | - | 3 | 402 | |||
AF534402.1 | - | 2 | 245 | |||
AK291663.1 | - | 3 | 368 | |||
AK292436.1 | - | 2 | 211 | |||
BC033640.1 | - | 2 | 206 | 5502950 |