Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281736.1 | NP_001268665.1 | 6 | 879 | ||
NM_006375.3 | NP_006366.2 | 7 | 1023 | |||
NM_182314.2 | NP_872114.1 | 8 | 1112 | |||
XM_005262354.3 | XP_005262411.1 | 7 | 968 | |||
XM_011531244.2 | XP_011529546.1 | 8 | 1114 | |||
XM_011531245.2 | XP_011529547.1 | 8 | 1114 | |||
XM_011531247.2 | XP_011529549.1 | 9 | 1240 | |||
XM_011531249.2 | XP_011529551.1 | 6 | 668 | |||
XM_011531251.2 | XP_011529553.1 | 6 | 773 | |||
XM_017029226.1 | XP_016884715.1 | 10 | 1331 | |||
XM_017029227.1 | XP_016884716.1 | 8 | 1065 | |||
XM_017029228.1 | XP_016884717.1 | 7 | 814 | |||
XM_017029229.1 | XP_016884718.1 | 7 | 1023 | |||
GenBank mRNA | AF207881.2 | - | 5 | 633 | ||
AK000353.1 | - | 6 | 796 | |||
AK289812.1 | - | 8 | 1004 | |||
AK289837.1 | - | 7 | 915 | |||
AL133207.2 | - | 7 | 910 | |||
BC019254.1 | - | 2 | 154 | |||
BC108713.1 | - | 7 | 900 | |||
BC139781.1 | - | 5 | 696 | |||
BC140874.1 | - | 6 | 793 | 9021742 |