Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005816.4 | NP_005807.1 | 14 | 4171 | ||
NM_198196.2 | NP_937839.1 | 15 | 4219 | |||
XM_006713469.3 | XP_006713532.1 | 15 | 4177 | |||
XM_006713470.3 | XP_006713533.1 | 14 | 4129 | |||
XM_017005521.1 | XP_016861010.1 | 14 | 3991 | |||
XM_017005522.1 | XP_016861011.1 | 12 | 3868 | |||
GenBank mRNA | AL833681.1 | - | 15 | 4066 |