Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF1 SIRNA Assays ›
Gene Symbol:
CELF1
Gene Name:
CUGBP, Elav-like family member 1
Gene Aliases:
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location:
Chr.11: 47465937 - 47565569 on Build GRCh38
UniGene ID:
Hs.595333
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137106
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025596.2
NP_001020767.1
6
666
NM_001172639.1
NP_001166110.1
9
1628
NM_001172640.1
NP_001166111.1
6
663
NM_006560.3
NP_006551.1
7
915
NM_198700.2
NP_941989.1
7
857
XM_011519848.2
XP_011518150.1
7
738
XM_011519850.2
XP_011518152.2
7
977
XM_011519852.2
XP_011518154.1
9
1086
XM_011519854.1
XP_011518156.1
11
1194
XM_011519855.1
XP_011518157.1
7
915
XM_011519856.1
XP_011518158.1
8
996
XM_011519857.1
XP_011518159.1
7
867
XM_011519858.1
XP_011518160.1
7
962
XM_011519859.1
XP_011518161.1
11
1194
XM_017017101.1
XP_016872590.1
10
1078
XM_017017102.1
XP_016872591.1
9
997
XM_017017103.1
XP_016872592.1
8
890
XM_017017104.1
XP_016872593.1
8
890
XM_017017105.1
XP_016872594.1
8
890
XM_017017106.1
XP_016872595.1
10
1078
XM_017017107.1
XP_016872596.1
8
890
XM_017017108.1
XP_016872597.1
8
890
XM_017017109.1
XP_016872598.1
7
977
XM_017017110.1
XP_016872599.1
8
890
XM_017017111.1
XP_016872600.1
8
890
XM_017017112.1
XP_016872601.1
8
890
XM_017017113.1
XP_016872602.1
8
890
XM_017017114.1
XP_016872603.1
8
890
XM_017017115.1
XP_016872604.1
9
918
XM_017017116.1
XP_016872605.1
8
953
XM_017017117.1
XP_016872606.1
7
714
XM_017017118.1
XP_016872607.1
7
836
XM_017017119.1
XP_016872608.1
9
1055
XM_017017120.1
XP_016872609.1
7
867
XM_017017121.1
XP_016872610.1
7
962
XM_017017122.1
XP_016872611.1
7
867
XM_017017123.1
XP_016872612.1
7
915
XM_017017124.1
XP_016872613.1
9
1009
XM_017017125.1
XP_016872614.1
7
738
XM_017017126.1
XP_016872615.1
7
867
XM_017017127.1
XP_016872616.1
7
891
XM_017017128.1
XP_016872617.1
7
915
XM_017017129.1
XP_016872618.1
8
913
XM_017017130.1
XP_016872619.1
7
867
XM_017017131.1
XP_016872620.1
7
891
XM_017017132.1
XP_016872621.1
7
915
XM_017017133.1
XP_016872622.1
7
706
XM_017017134.1
XP_016872623.1
8
845
XM_017017135.1
XP_016872624.1
7
867
GenBank mRNA
AB210019.1
-
9
1628
AF248648.1
-
6
674
AF267533.1
-
6
667
AF267534.1
-
6
667
AJ007988.1
-
6
656
AK304430.1
-
6
606
AK307679.1
-
1
128
BC031079.1
-
7
857
5297141
U63289.1
-
7
793
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splice site selection
mRNA processing
germ cell development
spermatid development
embryo development
RNA interference
negative regulation of translation
positive regulation of multicellular organism growth
regulation of RNA splicing
nucleotide binding
translation repressor activity, nucleic acid binding
RNA binding
mRNA binding
protein binding
translation initiation factor binding
BRE binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-sd57s:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline