Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 4, 5 | 2935 | ||
NM_001265590.1 | NP_001252519.1 | 3, 4 | 2599 | |||
NM_006054.3 | NP_006045.1 | 2, 3 | 547 | |||
NM_201428.2 | NP_958831.1 | 3, 4 | 2878 | |||
NM_201429.2 | NP_958832.1 | 3, 4 | 604 | |||
NM_201430.2 | NP_958833.1 | 2, 3 | 547 | |||
XM_011544730.2 | XP_011543032.1 | 4, 5 | 3025 | |||
XM_011544731.2 | XP_011543033.1 | 3, 4 | 739 | |||
GenBank mRNA | AB209771.1 | - | 2, 3 | 484 | ||
AF059524.1 | - | 2, 3 | 484 | |||
AF119297.1 | - | 2, 3 | 496 | |||
AK075412.1 | - | 2, 3 | 497 | |||
AK094965.1 | - | 2, 3 | 1600 | |||
AK122727.1 | - | 2, 3 | 1000 | |||
AK127079.1 | - | 3, 4 | 2574 | |||
AK222898.1 | - | 2, 3 | 484 | |||
AK295361.1 | - | 3, 4 | 509 | |||
AK295534.1 | - | 2, 3 | 401 | |||
AK295758.1 | - | 3, 4 | 2527 | |||
AK304100.1 | - | 4, 5 | 2885 | |||
AK307811.1 | - | 3, 4 | 2824 | |||
AK308083.1 | - | 2, 3 | 492 | |||
AK309766.1 | - | 3, 4 | 2553 | |||
AY427821.2 | - | 3, 4 | 2845 | |||
AY750848.1 | - | 4, 5 | 2902 | |||
BC000634.1 | - | 2, 3 | 455 | |||
BC010556.1 | - | 2, 3 | 460 | |||
BC011394.1 | - | 2, 3 | 496 | |||
BC022993.1 | - | 2, 3 | 486 | |||
BC100822.2 | - | 2, 3 | 477 | |||
BC100823.2 | - | 2, 3 | 477 | |||
BC105981.1 | - | 2, 3 | 428 | |||
BC105982.1 | - | 2, 3 | 459 | |||
BC118550.1 | - | 2, 3 | 477 | |||
BC118628.1 | - | 2, 3 | 477 |