Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 9 | 4469 | ||
NM_001265590.1 | NP_001252519.1 | 8 | 4133 | |||
NM_001265591.1 | NP_001252520.1 | 4 | 1825 | |||
NM_006054.3 | NP_006045.1 | 7 | 2081 | |||
NM_201428.2 | NP_958831.1 | 8 | 4412 | |||
NM_201429.2 | NP_958832.1 | 8 | 2138 | |||
NM_201430.2 | NP_958833.1 | 6 | 2022 | |||
NR_049750.1 | 6 | 1942 | ||||
NR_049751.1 | 5 | 1883 | ||||
XM_011544730.2 | XP_011543032.1 | 8 | 4500 | |||
XM_011544731.2 | XP_011543033.1 | 7 | 2214 | |||
GenBank mRNA | AF059524.1 | - | 7 | 2026 | ||
AF119297.1 | - | 7 | 2030 | |||
AK075412.1 | - | 7 | 2030 | |||
AK094965.1 | - | 7 | 3135 | |||
AK122727.1 | - | 7 | 2533 | |||
AK127079.1 | - | 8 | 4107 | |||
AY427821.2 | - | 8 | 4379 | |||
AY750848.1 | - | 9 | 4436 | |||
BC000865.1 | - | 1 | 40 | |||
BC011394.1 | - | 7 | 2030 | |||
BC022993.1 | - | 7 | 2020 | |||
BC100822.2 | - | 7 | 2011 | |||
BC100823.2 | - | 7 | 2011 | |||
BC105981.1 | - | 7 | 1962 | |||
BC105982.1 | - | 7 | 1993 | |||
BC118550.1 | - | 7 | 2011 | |||
BC118628.1 | - | 7 | 2011 |