Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 6 | 3120 | ||
NM_001265590.1 | NP_001252519.1 | 5 | 2784 | |||
NM_006054.3 | NP_006045.1 | 4 | 732 | |||
NM_201428.2 | NP_958831.1 | 5 | 3063 | |||
NM_201429.2 | NP_958832.1 | 5 | 789 | |||
NM_201430.2 | NP_958833.1 | 4 | 732 | |||
NR_049750.1 | 3 | 593 | ||||
NR_049751.1 | 3 | 593 | ||||
XM_011544730.2 | XP_011543032.1 | 6 | 3210 | |||
XM_011544731.2 | XP_011543033.1 | 5 | 924 | |||
GenBank mRNA | AB209771.1 | - | 4 | 669 | ||
AF059524.1 | - | 4 | 669 | |||
AF119297.1 | - | 4 | 681 | |||
AK075412.1 | - | 4 | 682 | |||
AK094965.1 | - | 4 | 1785 | |||
AK122727.1 | - | 4 | 1185 | |||
AK127079.1 | - | 5 | 2759 | |||
AK222898.1 | - | 4 | 669 | |||
AK295361.1 | - | 5 | 694 | |||
AK295534.1 | - | 4 | 586 | |||
AK295758.1 | - | 5 | 2712 | |||
AK297533.1 | - | 3 | 543 | |||
AK304100.1 | - | 6 | 3070 | |||
AK307811.1 | - | 5 | 3009 | |||
AK308083.1 | - | 4 | 677 | |||
AK309766.1 | - | 5 | 2738 | |||
AY427821.2 | - | 5 | 3030 | |||
AY750848.1 | - | 6 | 3087 | |||
BC000634.1 | - | 4 | 640 | |||
BC010556.1 | - | 4 | 645 | |||
BC011394.1 | - | 4 | 681 | |||
BC022993.1 | - | 4 | 671 | |||
BC100822.2 | - | 4 | 662 | |||
BC100823.2 | - | 4 | 662 | |||
BC105981.1 | - | 4 | 613 | |||
BC105982.1 | - | 4 | 644 | |||
BC118550.1 | - | 4 | 662 | |||
BC118628.1 | - | 4 | 662 |