Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013436.4 | NP_038464.1 | 13 | 2050 | ||
NM_205842.2 | NP_995314.1 | 14 | 2068 | |||
XM_006712200.3 | XP_006712263.1 | 14 | 1554 | |||
XM_006712201.3 | XP_006712264.1 | 13 | 1538 | |||
GenBank mRNA | AB011159.1 | - | 14 | 2068 | ||
AB014509.1 | - | 13 | 1310 | |||
AK292914.1 | - | 13 | 1540 | |||
BC015025.1 | - | 13 | 1562 | 3907646 | ||
BT007033.1 | - | 13 | 1291 |