Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083.3 | NP_001074.2 | 2, 3 | 904 | ||
NM_033430.2 | NP_236914.2 | 2, 3 | 735 | |||
NM_033437.3 | NP_246273.2 | 2, 3 | 687 | |||
XM_017008791.1 | XP_016864280.1 | 2, 3 | 1158 | |||
GenBank mRNA | AB015656.1 | - | 2, 3 | 629 | ||
AF043731.1 | - | 2, 3 | 904 | |||
AJ004865.1 | - | 2, 3 | 842 | |||
AY264918.1 | - | 2, 3 | 623 | |||
AY266363.1 | - | 1, 2 | 283 | |||
BC126233.1 | - | 2, 3 | 838 | 8991949 | ||
CD511199.1 | - | 2, 3 | 652 | |||
JQ286346.1 | - | 2, 3 | 749 |