Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003737.3 | NP_003728.1 | 2 | 1922 | ||
GenBank mRNA | AB000895.1 | - | 1 | 216 | ||
AB053446.1 | - | 2 | 1925 |