Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161504.1 | NP_001154976.1 | 10 | 1257 | ||
NM_001319218.1 | NP_001306147.1 | 10 | 1397 | |||
NM_003748.3 | NP_003739.2 | 10 | 1397 | |||
NM_170726.2 | NP_733844.1 | 10 | 1397 | |||
GenBank mRNA | AK289972.1 | - | 10 | 1323 | ||
AK294552.1 | - | 10 | 1257 | |||
BC007581.1 | - | 10 | 1127 | 3139944 | ||
BC023600.2 | - | 10 | 1138 | 4548787 | ||
FJ462711.1 | - | 10 | 1135 | |||
U24266.1 | - | 10 | 1151 | |||
U24267.1 | - | 10 | 1151 |