Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037283.1 | NP_001032360.1 | 10 | 1657 | ||
NM_003751.3 | NP_003742.2 | 10 | 1657 | |||
XM_011515599.1 | XP_011513901.1 | 10 | 1956 | |||
XM_011515600.1 | XP_011513902.1 | 10 | 1956 | |||
XM_011515601.1 | XP_011513903.1 | 11 | 1839 | |||
XM_017012752.1 | XP_016868241.1 | 10 | 1956 | |||
GenBank mRNA | AB209381.1 | - | 3 | 3207 | ||
AK300967.1 | - | 11 | 1399 | |||
AK302058.1 | - | 11 | 1183 | |||
BC001173.1 | - | 10 | 1605 | 3501759 | ||
BC009986.1 | - | 9 | 999 | |||
BC110865.1 | - | 10 | 1583 | 6175284 | ||
U62583.1 | - | 10 | 1627 | |||
U78525.1 | - | 10 | 1649 |