Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252641.1 | NP_001239570.1 | 7 | 527 | ||
NM_003796.3 | NP_003787.2 | 7 | 842 | |||
NR_045557.1 | 6 | 807 | ||||
XM_005259362.2 | XP_005259419.1 | 7 | 521 | |||
XM_005259363.4 | XP_005259420.1 | 7 | 578 | |||
XM_011527435.1 | XP_011525737.1 | 6 | 404 | |||
GenBank mRNA | AF091095.1 | - | 8 | 478 | ||
AK001437.1 | - | 6 | 528 | |||
AK226173.1 | - | 7 | 830 | |||
AK292170.1 | - | 7 | 578 | |||
AK314313.1 | - | 7 | 567 | |||
BC014933.2 | - | 2 | 59 | |||
BC026184.2 | - | 7 | 527 | |||
BC067259.1 | - | 6 | 534 | 6500727 |