Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317061.1 | NP_001303990.1 | 11 | 2041 | ||
NM_003804.4 | NP_003795.2 | 9 | 1763 | |||
XM_005249457.3 | XP_005249514.2 | 9 | 1686 | |||
XM_017011403.1 | XP_016866892.1 | 6 | 1795 | |||
XM_017011404.1 | XP_016866893.1 | 3 | 1003 | |||
GenBank mRNA | AB208926.1 | - | 11 | 1950 | ||
AK096523.1 | - | 3 | 531 | |||
AK304701.1 | - | 7 | 1352 | |||
AK314176.1 | - | 9 | 1763 | |||
BC126254.1 | - | 8 | 1557 | 8991970 | ||
BC126256.1 | - | 8 | 1557 | 8991972 | ||
BX348786.2 | - | 2 | 614 | |||
U50062.1 | - | 8 | 1531 |