Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306144.1 | NP_001293073.1 | 10 | 1097 | ||
NM_001306145.1 | NP_001293074.1 | 9 | 1073 | |||
NM_003828.3 | NP_003819.1 | 9 | 1073 | |||
XM_005274765.3 | XP_005274822.1 | 12 | 1137 | |||
XM_005274766.3 | XP_005274823.1 | 11 | 1085 | |||
XM_005274767.3 | XP_005274824.1 | 10 | 1104 | |||
XM_006724855.3 | XP_006724918.1 | 9 | 1073 | |||
XM_011531209.2 | XP_011529511.1 | 9 | 833 | |||
XM_011531210.2 | XP_011529512.1 | 12 | 1139 | |||
XM_017029920.1 | XP_016885409.1 | 12 | 1137 | |||
XM_017029921.1 | XP_016885410.1 | 10 | 868 | |||
XM_017029922.1 | XP_016885411.1 | 10 | 880 | |||
XM_017029923.1 | XP_016885412.1 | 7 | 660 | |||
XM_017029924.1 | XP_016885413.1 | 10 | 1104 | |||
GenBank mRNA | AF057354.1 | - | 9 | 897 | ||
AJ224979.1 | - | 9 | 899 | |||
AK001816.1 | - | 7 | 602 | |||
AK226109.1 | - | 10 | 1051 | |||
AK295738.1 | - | 10 | 892 | |||
BC032947.1 | - | 9 | 1070 | 4837744 | ||
U58032.1 | - | 6 | 678 |