Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003830.3 | NP_003821.1 | 5 | 1047 | ||
XM_011527438.2 | XP_011525740.1 | 5 | 1001 | |||
XM_017027419.1 | XP_016882908.1 | 5 | 1001 | |||
GenBank mRNA | AF170484.1 | - | 5 | 944 | ||
AK225753.1 | - | 5 | 968 | |||
BC029896.1 | - | 5 | 949 | 5178889 | ||
U71383.1 | - | 5 | 1050 |