Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003837.3 | NP_003828.2 | 7 | 1294 | ||
GenBank mRNA | AJ573829.1 | - | 1 | 366 | ||
Y10812.2 | - | 7 | 1255 |