Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003845.2 | NP_003836.1 | 7, 8 | 870 | ||
GenBank mRNA | AF263541.1 | - | 6, 7 | 770 | ||
AK292322.1 | - | 7, 8 | 864 | |||
AK308260.1 | - | 9, 10 | 1091 | |||
BC031244.1 | - | 7, 8 | 864 | 5266287 | ||
GU139743.1 | - | 6, 7 | 854 |