Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271572.1 | NP_001258501.1 | 3 | 796 | ||
NM_001271573.1 | NP_001258502.1 | 2 | 773 | |||
NM_003847.2 | NP_003838.1 | 3 | 889 | |||
GenBank mRNA | AB015594.1 | - | 3 | 832 | ||
AF093668.1 | - | 3 | 794 | |||
AK001415.1 | - | 3 | 882 | |||
AK297442.1 | - | 2 | 621 | |||
AK300978.1 | - | 3 | 674 | |||
BC009697.1 | - | 3 | 783 | 3878406 |