Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003851.2 | NP_003842.1 | 4 | 1102 | ||
GenBank mRNA | AF084523.1 | - | 4 | 1057 | ||
AK075375.1 | - | 4 | 1063 | |||
AK095456.1 | - | 5 | 1021 | |||
AK130006.1 | - | 5 | 949 | |||
BC006786.1 | - | 4 | 1033 | 2900817 | ||
BC008628.1 | - | 4 | 1033 | 4339480 |