Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135191.1 | NP_001128663.1 | 6 | 843 | ||
NM_003887.2 | NP_003878.1 | 6 | 843 | |||
XM_006711898.1 | XP_006711961.1 | 6 | 596 | |||
XM_006711901.3 | XP_006711964.1 | 6 | 446 | |||
XM_006711902.1 | XP_006711965.1 | 6 | 596 | |||
XM_011510403.2 | XP_011508705.1 | 6 | 617 | |||
XM_011510404.2 | XP_011508706.1 | 6 | 616 | |||
XM_011510405.2 | XP_011508707.1 | 5 | 543 | |||
XM_011510406.2 | XP_011508708.1 | 7 | 550 | |||
XM_011510407.2 | XP_011508709.1 | 6 | 617 | |||
XM_011510408.2 | XP_011508710.1 | 6 | 620 | |||
XM_011510409.1 | XP_011508711.1 | 6 | 488 | |||
GenBank mRNA | AB007860.1 | - | 6 | 843 | ||
BC063308.1 | - | 6 | 796 | 30345039 |