Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135191.1 | NP_001128663.1 | 27 | 5441 | ||
NM_003887.2 | NP_003878.1 | 28 | 5576 | |||
XM_006711898.1 | XP_006711961.1 | 29 | 5338 | |||
XM_006711901.3 | XP_006711964.1 | 29 | 5188 | |||
XM_006711902.1 | XP_006711965.1 | 28 | 5203 | |||
XM_011510403.2 | XP_011508705.1 | 29 | 5359 | |||
XM_011510404.2 | XP_011508706.1 | 28 | 5349 | |||
XM_011510405.2 | XP_011508707.1 | 28 | 5285 | |||
XM_011510406.2 | XP_011508708.1 | 30 | 5292 | |||
XM_011510407.2 | XP_011508709.1 | 28 | 5224 | |||
XM_011510409.1 | XP_011508711.1 | 29 | 5230 | |||
GenBank mRNA | AB007860.1 | - | 28 | 5576 | ||
AK091121.1 | - | 6 | 2988 | |||
AK124900.1 | - | 4 | 3735 | |||
BC063308.1 | - | 27 | 5393 | 30345039 |