Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206897.1 | NP_001193826.1 | 10 | 1380 | ||
NM_003888.3 | NP_003879.2 | 9 | 1283 | |||
NM_170696.2 | NP_733797.1 | 8 | 1169 | |||
NM_170697.2 | NP_733798.1 | 7 | 887 | |||
GenBank mRNA | AB015226.1 | - | 9 | 1053 | ||
AB015227.1 | - | 7 | 816 | |||
AB015228.1 | - | 7 | 778 | |||
AK128709.1 | - | 9 | 1778 | |||
AK294981.1 | - | 9 | 983 | |||
AK303057.1 | - | 10 | 1125 | |||
AL110299.1 | - | 1 | 831 | |||
BC030589.2 | - | 8 | 927 | 4826743 |