Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078645.1 | NP_001072113.1 | 8 | 848 | ||
NM_001318517.1 | NP_001305446.1 | 8 | 845 | |||
NM_001318518.1 | NP_001305447.1 | 8 | 845 | |||
NM_003903.3 | NP_003894.3 | 8 | 848 | |||
XM_005266206.1 | XP_005266263.1 | 8 | 890 | |||
XM_006719992.1 | XP_006720055.1 | 8 | 835 | |||
XM_006719993.1 | XP_006720056.1 | 7 | 783 | |||
XM_011534851.1 | XP_011533153.1 | 8 | 890 | |||
XM_011534853.1 | XP_011533155.1 | 8 | 890 | |||
XM_017020830.1 | XP_016876319.1 | 8 | 890 | |||
XM_017020831.1 | XP_016876320.1 | 8 | 890 | |||
XM_017020832.1 | XP_016876321.1 | 7 | 783 | |||
XM_017020833.1 | XP_016876322.1 | 6 | 468 | |||
GenBank mRNA | AB209850.1 | - | 6 | 2977 | ||
AF164598.1 | - | 8 | 651 | |||
AK315302.1 | - | 8 | 679 | |||
BC010875.2 | - | 8 | 792 | 3879150 | ||
BC017244.1 | - | 8 | 758 | 4877392 | ||
U18291.1 | - | 9 | 671 |