Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078645.1 | NP_001072113.1 | 13 | 1348 | ||
NM_001318517.1 | NP_001305446.1 | 13 | 1345 | |||
NM_003903.3 | NP_003894.3 | 13 | 1348 | |||
XM_005266206.1 | XP_005266263.1 | 13 | 1390 | |||
XM_006719992.1 | XP_006720055.1 | 13 | 1335 | |||
XM_006719993.1 | XP_006720056.1 | 12 | 1283 | |||
XM_011534853.1 | XP_011533155.1 | 13 | 1390 | |||
XM_017020831.1 | XP_016876320.1 | 13 | 1390 | |||
XM_017020832.1 | XP_016876321.1 | 12 | 1283 | |||
XM_017020833.1 | XP_016876322.1 | 11 | 968 | |||
GenBank mRNA | AB209850.1 | - | 10 | 4377 | ||
AK315302.1 | - | 13 | 1179 | |||
BC010875.2 | - | 13 | 1292 | 3879150 | ||
BC017244.1 | - | 13 | 1258 | 4877392 | ||
U18291.1 | - | 14 | 1171 |