Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018159.1 | NP_001018169.1 | 5, 6 | 419 | ||
NM_001018160.1 | NP_001018170.1 | 3, 4 | 217 | |||
NM_001286500.1 | NP_001273429.1 | 4, 5 | 330 | |||
NM_003905.3 | NP_003896.1 | 4, 5 | 321 | |||
GenBank mRNA | AK097680.1 | - | 4, 5 | 330 | ||
AK298159.1 | - | 5, 6 | 368 | |||
AK307677.1 | - | 3, 4 | 209 | |||
AK312784.1 | - | 4, 5 | 321 | |||
AL136798.1 | - | 4, 5 | 290 | |||
AY197612.1 | - | 4, 5 | 259 | |||
BC000480.2 | - | 4, 5 | 252 | 2964638 | ||
BC013301.2 | - | 4, 5 | 252 | 2964638 | ||
BM045535.1 | - | 5, 6 | 390 | |||
BM785899.1 | - | 4, 5 | 321 | |||
U50939.1 | - | 4, 5 | 317 |