Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM3AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003906.4 | NP_003897.2 | 1 | 262 | ||
XM_005261203.4 | XP_005261260.1 | 2 | 838 | |||
XM_005261204.4 | XP_005261261.1 | 2 | 734 | |||
XM_005261205.3 | XP_005261262.1 | 2 | 350 | |||
GenBank mRNA | AJ010089.1 | - | 1 | 191 | ||
BC104958.1 | - | 1 | 230 | 8143961 | ||
BC104960.1 | - | 1 | 230 | 8143963 | ||
HF584748.1 | - | 2 | 320 |